Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028331.2 | 2 | 220 | |||
GenBank mRNA | AK008471.1 | - | 2 | 196 | ||
AK019676.1 | - | 2 | 170 | |||
AK029711.1 | - | 2 | 388 | |||
AK085166.1 | - | 2, 3 | 180 | |||
BB615378.1 | - | 2 | 381 | |||
BC034706.1 | - | 2 | 167 | 3582315 | ||
BU148968.1 | - | 2 | 152 | |||
BY017375.1 | - | 2, 3 | 201 | |||
BY715308.1 | - | 2 | 170 | |||
CA321162.1 | - | 2 | 153 | |||
CV783882.1 | - | 2 | 169 | |||
DV654279.1 | - | 2 | 255 |