Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bambi-ps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027919.1 | 1 | 507 | |||
GenBank mRNA | AK040009.1 | - | 1 | 451 | ||
AK041875.1 | - | 1 | 453 | |||
AK088217.1 | - | 1 | 445 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BF720956.1 | - | 1 | 507 | Chr. 2 - 122467275 - 122467797 [-] on Build GRCm38 |