Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP6V0E2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027040.1 | 4 | 1889 | |||
GenBank mRNA | BC023225.1 | - | 4 | 1702 | ||
BC028735.2 | - | 3 | 1759 | |||
BC092426.1 | - | 1 | 499 |