Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2IRD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003934.1 | 10, 11 | 876 | |||
GenBank mRNA | AK093373.1 | - | 10, 11 | 876 |