Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021487.2 | 6 | 3763 | |||
NR_027328.1 | 4 | 5604 | ||||
NR_027329.1 | 5 | 5712 | ||||
GenBank mRNA | AB067495.1 | - | 4 | 5582 | ||
AK091044.1 | - | 1 | 2959 | |||
AK092212.1 | - | 1 | 2206 | |||
BC036405.1 | - | 6 | 3571 | |||
BC047649.1 | - | 6 | 3566 |