Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMG3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021487.2 | 5 | 1364 | |||
NR_027328.1 | 4 | 1002 | ||||
NR_027329.1 | 5 | 1110 | ||||
GenBank mRNA | AB067495.1 | - | 4 | 980 | ||
AK054665.1 | - | 4 | 1002 | |||
BC036405.1 | - | 5 | 1172 | |||
BC047649.1 | - | 5 | 1167 | |||
BX538028.1 | - | 5 | 1048 |