Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAHD2CP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003698.1 | 4 | 801 | |||
GenBank mRNA | BI916267.1 | - | 4 | 767 |
Gene Symbol: FAHD2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011510745.2 | XP_011509047.1 | 10 | 1458 | ||
XM_011510746.2 | XP_011509048.1 | 10 | 1577 | |||
XM_011510747.2 | XP_011509049.1 | 9 | 1342 | |||
XM_011510748.2 | XP_011509050.1 | 10 | 1422 | |||
XM_017003470.1 | XP_016858959.1 | 10 | 1455 | |||
XM_017003471.1 | XP_016858960.1 | 10 | 1360 | |||
XM_017003472.1 | XP_016858961.1 | 9 | 1391 |
Gene Symbol: LOC653924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC104425.1 | - | 1 | 321 |