Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 6 | 839 | |||
NR_027626.1 | 7 | 1158 | ||||
GenBank mRNA | AF074480.1 | - | 7 | 859 | ||
AK303297.1 | - | 7 | 1115 | |||
BC032500.1 | - | 7 | 1051 | |||
CR749466.1 | - | 5 | 642 | |||
FJ794466.1 | - | 6 | 817 |