Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX12P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002710.2 | 11 | 1920 | |||
NR_120453.1 | 10 | 1367 | ||||
GenBank mRNA | AL832768.1 | - | 11 | 1637 | ||
BC041851.1 | - | 13 | 2101 |