Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALOX12P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002710.2 | 1 | 332 | |||
GenBank mRNA | AL832768.1 | - | 1 | 49 | ||
BC041851.1 | - | 1 | 332 |