Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003040.2 | 1 | 577 | |||
GenBank mRNA | BC017930.1 | - | 1 | 581 | ||
BC060042.1 | - | 1 | 584 |