Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024458.1 | 7 | 685 | |||
GenBank mRNA | AF318337.1 | - | 7 | 685 | ||
AK124332.1 | - | 5 | 600 |