Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBL1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002724.2 | 1 | 133 | |||
GenBank mRNA | AF019382.1 | - | LIT2047 MBL1P1 |
1 | 132 |