Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028412.1 | 3 | 722 | |||
NR_028413.1 | 3 | 537 | ||||
NR_028414.1 | 4 | 414 | ||||
GenBank mRNA | AV645786.1 | - | 3 | 575 | ||
CB162526.1 | - | 3 | 499 | |||
DA636997.1 | - | 4 | 413 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV662258.1 | - | 257 | Chr. - - [-] on Build |