Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOC1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028412.1 | 2 | 512 | |||
NR_028413.1 | 2 | 327 | ||||
NR_028414.1 | 3 | 204 | ||||
GenBank mRNA | AV645786.1 | - | 2 | 365 | ||
CB162526.1 | - | 2 | 289 | |||
DA636997.1 | - | 3 | 203 | |||
DA638554.1 | - | 2 | 327 | |||
EF553526.1 | - | 2 | 327 |