Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPM3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003148.3 | 2 | 2719 | |||
GenBank mRNA | AK094777.1 | - | 4 | 2660 | ||
BX647962.1 | - | 2 | 2705 |
Gene Symbol: KLHL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC037989.1 | - | 4 | 2345 |
Gene Symbol: ZNF549 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AL833090.2 | - | 4 | 5358 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BC040666.1 | - | 1 | 3822 | Chr. 14 - 103525006 - 103529041 [-] on Build GRCh38 |