Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNM1P46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003260.1 | 5 | 2805 | |||
GenBank mRNA | AK127834.1 | - | 5 | 2805 | ||
AK302717.1 | - | 5 | 2606 | |||
AL833912.1 | - | 4 | 2660 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125787.1 | - | 5 | 3428 | Chr. 15 - 99790781 - 99801688 [-] on Build GRCh38 |