Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF503-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024421.1 | 2 | 1350 | |||
NR_024422.1 | 2 | 918 | ||||
NR_110299.1 | 2 | 914 | ||||
NR_110300.1 | 2 | 686 | ||||
NR_110301.1 | 2 | 803 | ||||
NR_110302.1 | 2 | 736 | ||||
GenBank mRNA | BC029963.1 | - | 2 | 686 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX344557.2 | - | 2 | 979 | Chr. 10 - 75401898 - 75407941 [+] on Build GRCh38 |