Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317951.1 | NP_001304880.1 | 7 | 1059 | ||
XM_006714494.3 | XP_006714557.1 | 7 | 919 | |||
XM_017009780.1 | XP_016865269.1 | 7 | 919 | |||
GenBank mRNA | AK023503.1 | - | 7 | 1037 |