Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317951.1 | NP_001304880.1 | 4 | 603 | ||
NR_134296.1 | 4 | 820 | ||||
GenBank mRNA | AK023503.1 | - | 4 | 581 | ||
AK024392.1 | - | 1 | 319 | |||
AK301987.1 | - | 4 | 435 | |||
BC007217.1 | - | 3 | 327 | |||
BC041590.1 | - | 2 | 170 | 5428011 | ||
BX648703.1 | - | 4 | 822 |