Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026052.1 | 2 | 3098 | |||
GenBank mRNA | AA418278.1 | - | 1 | 173 | ||
AK098293.1 | - | 1 | 2103 | |||
BC043401.1 | - | 2 | 3037 | |||
BC098570.1 | - | 1 | 2608 |