Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPBD1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026052.1 | 2 | 2116 | |||
GenBank mRNA | AK098293.1 | - | 1 | 1121 | ||
BC043401.1 | - | 2 | 2055 | |||
BC098570.1 | - | 1 | 1626 | |||
BC119703.1 | - | 1 | 1487 |