Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CST13P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001279.2 | 4 | 984 | |||
GenBank mRNA | BC024006.1 | - | 4 | 989 | ||
BC028368.1 | - | 4 | 989 | |||
BC033934.1 | - | 4 | 979 | |||
BC041171.1 | - | 4 | 963 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB522378.1 | - | 3 | 300 | Chr. 20 - 23521092 - 23542012 [+] on Build GRCh38 |