Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276318.1 | NP_001263247.1 | 5 | 2671 | ||
GenBank mRNA | AK024489.1 | - | 5 | 2671 | ||
AK091943.1 | - | 5 | 983 |