Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BACH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027655.2 | 5 | 2123 | |||
GenBank mRNA | AF317902.1 | - | 1 | 168 | ||
DB524401.1 | - | 1 | 39 |