Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027271.1 | 1 | 416 | |||
GenBank mRNA | BC026041.1 | - | 1 | 415 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY158516.1 | - | 1 | 211 | Chr. 19 - 1268099 - 1270054 [-] on Build GRCh38 |