Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NBEAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027992.1 | 2 | 247 | |||
GenBank mRNA | AL832227.1 | - | 2 | 385 | ||
DA278599.1 | - | 2 | 247 |