Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR9-3HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015411.1 | 7 | 4177 | |||
NR_133001.1 | 7 | 4757 | ||||
NR_133002.1 | 6 | 4058 | ||||
NR_133003.1 | 7 | 4092 | ||||
GenBank mRNA | AK054710.1 | - | 1 | 1793 | ||
AK054931.1 | - | 8 | 2103 | |||
AK096359.1 | - | 1 | 2292 |