Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARVELD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AI347102.1 | - | 1 | 145 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY250975.1 | - | 1 | 152 | Chr. 10 - 97717708 - 97718144 [-] on Build GRCh38 |