Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DLEU2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002612.1 | 11 | 1698 | |||
GenBank mRNA | AF529010.1 | - | 4 | 715 | ||
AY455757.1 | - | LIT3420 DLEU2 |
14 | 1998 |