Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC90768 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027107.2 | 4 | 4797 | |||
NR_125905.1 | 5 | 2844 | ||||
GenBank mRNA | AK094166.1 | - | 1 | 2301 |