Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102724958 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110760.1 | 1 | 124 | |||
GenBank mRNA | AY726575.1 | - | 1 | 124 |
Gene Symbol: LINC00906 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027318.2 | 2 | 2887 | |||
GenBank mRNA | BX647231.1 | - | 2 | 2885 | ||
DB513590.2 | - | 1 | 142 |