Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026983.1 | 1 | 223 | |||
GenBank mRNA | BC106717.2 | - | 1 | 608 | ||
BC106718.2 | - | 1 | 608 | |||
BC115358.1 | - | 1 | 608 |