Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3P11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026983.1 | 1 | 599 |
Gene Symbol: BTF3P9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC062736.1 | - | 1 | 841 |