Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC5A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028323.1 | 11 | 1813 | |||
GenBank mRNA | AK027855.1 | - | 11 | 1654 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA342747.1 | - | 1 | 207 | Chr. 2 - 27202022 - 27203328 [-] on Build GRCh38 |