Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243571.1 | NP_001230500.1 | 4 | 574 | ||
XM_017018517.1 | XP_016874006.1 | 3 | 440 |