Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00905 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024335.2 | 2 | 437 | |||
NR_110321.1 | 2 | 147 | ||||
GenBank mRNA | AK058057.1 | - | 2 | 396 | ||
BC031284.1 | - | 2 | 147 | |||
BC069223.1 | - | 2 | 369 | |||
BC104221.2 | - | 1 | 334 | |||
DB461577.1 | - | 2 | 437 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI562021.1 | - | 2 | 152 | Chr. 19 - 16034120 - 16041522 [+] on Build GRCh38 |