Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC13A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027384.1 | 2 | 192 | |||
XM_006722165.2 | XP_006722228.1 | 2 | 100 | |||
XM_011525451.1 | XP_011523753.1 | 2 | 113 | |||
GenBank mRNA | AK091926.1 | - | 2 | 100 | ||
AK303909.1 | - | 2 | 100 | |||
BC096276.1 | - | 2 | 146 | 40006564 |