Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EFHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033327.1 | 4 | 1968 | |||
GenBank mRNA | AK225543.1 | - | 4 | 1860 | ||
AL122084.1 | - | 4 | 1855 |