Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC148413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015434.1 | 3 | 1735 | |||
GenBank mRNA | BC068600.1 | - | 3 | 1692 | ||
BC080546.1 | - | 3 | 1691 |
Gene Symbol: MRPL20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AA824533.1 | - | 1 | 143 | ||
AW015403.1 | - | 1 | 154 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY250895.1 | - | 1 | 156 | Chr. 1 - 1401909 - 1406964 [+] on Build GRCh38 |