Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023312.2 | 6 | 2595 | |||
NR_023313.2 | 5 | 2427 | ||||
XM_011527668.1 | XP_011525970.1 | 7 | 1028 | |||
GenBank mRNA | AK094781.1 | - | 5 | 2361 | ||
AK095781.1 | - | 4 | 2654 | |||
AK128423.1 | - | 5 | 2448 | |||
AL133636.1 | - | 1 | 804 | |||
BI819825.1 | - | 1 | 547 |