Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIRBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300829.1 | NP_001287758.1 | 6 | 787 | ||
NR_023312.2 | 6 | 846 | ||||
NR_023313.2 | 5 | 678 | ||||
GenBank mRNA | AK094781.1 | - | 5 | 611 | ||
AK095781.1 | - | 4 | 906 | |||
AK128423.1 | - | 5 | 700 | |||
AK304185.1 | - | 5 | 864 | |||
AK304470.1 | - | 6 | 619 | |||
AK311601.1 | - | 5 | 1088 |