Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 4 | 392 | |||
GenBank mRNA | AK097598.1 | - | 2 | 153 | ||
BC048276.1 | - | 4 | 392 |
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 4 | 393 | |||
NR_110481.1 | 2 | 125 | ||||
GenBank mRNA | AK058056.1 | - | 4 | 393 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546201.1 | XP_011544503.1 | 2 | 152 | ||
XM_011546205.1 | XP_011544507.1 | 6 | 563 | |||
XM_011546206.1 | XP_011544508.1 | 5 | 540 | |||
XM_011546222.1 | XP_011544524.1 | 1 | 40 |