Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 8 | 1092 | |||
GenBank mRNA | AK097598.1 | - | 6 | 853 | ||
BC048276.1 | - | 8 | 1092 |
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 7 | 704 | |||
NR_110481.1 | 5 | 436 | ||||
GenBank mRNA | AK058056.1 | - | 7 | 704 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546201.1 | XP_011544503.1 | 5 | 463 | ||
XM_011546205.1 | XP_011544507.1 | 9 | 874 | |||
XM_011546206.1 | XP_011544508.1 | 8 | 851 |