Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 6 | 847 | |||
GenBank mRNA | AK097598.1 | - | 4 | 608 | ||
BC048276.1 | - | 6 | 847 |
Gene Symbol: LOC101927079 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110480.1 | 5 | 459 | |||
NR_110481.1 | 3 | 191 | ||||
GenBank mRNA | AK058056.1 | - | 5 | 459 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011546201.1 | XP_011544503.1 | 3 | 218 | ||
XM_011546205.1 | XP_011544507.1 | 7 | 629 | |||
XM_011546206.1 | XP_011544508.1 | 6 | 606 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY026677.1 | - | 5 | 460 | Chr. 15|NT_187382.1 - 279882 - 350572 [+] on Build GRCh38 |