Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC727924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015416.1 | 10, 11 | 1507 | |||
GenBank mRNA | BC048276.1 | - | 10, 11 | 1507 |
Gene Symbol: LOC102723532 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030118.1 | XP_016885607.1 | 2, 3 | 285 |