Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026590.1 | 1 | 48 | |||
GenBank mRNA | AF081258.1 | - | 1 | 48 | ||
AF081259.1 | - | 1 | 48 |