Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301251.1 | NP_001288180.1 | 2, 3 | 245 | ||
NM_001301252.1 | NP_001288181.1 | 1, 2 | 559 | |||
NM_001301253.1 | NP_001288182.1 | 2, 3 | 235 | |||
NM_001301254.1 | NP_001288183.1 | 1, 2 | 559 | |||
XM_017016900.1 | XP_016872389.1 | 2, 3 | 275 | |||
XM_017016901.1 | XP_016872390.1 | 2, 3 | 255 | |||
XM_017016902.1 | XP_016872391.1 | 2, 3 | 274 | |||
XM_017016903.1 | XP_016872392.1 | 2, 3 | 259 | |||
XM_017016904.1 | XP_016872393.1 | 2, 3 | 287 | |||
XM_017016905.1 | XP_016872394.1 | 2, 3 | 273 | |||
XM_017016907.1 | XP_016872396.1 | 2, 3 | 287 | |||
XM_017016908.1 | XP_016872397.1 | 2, 3 | 276 | |||
GenBank mRNA | AB082527.1 | - | 2, 3 | 194 | ||
AF505653.1 | - | 2, 3 | 232 | |||
AK294510.1 | - | 1, 2 | 477 | |||
AK304842.1 | - | 1, 2 | 210 |