Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027941.2 | 4 | 955 | |||
NR_027942.2 | 4 | 941 | ||||
NR_135677.1 | 4 | 2544 | ||||
XM_011524342.2 | XP_011522644.1 | 4 | 4783 | |||
XM_017024192.1 | XP_016879681.1 | 4 | 4783 | |||
XM_017024193.1 | XP_016879682.1 | 4 | 4783 | |||
XM_017024194.1 | XP_016879683.1 | 4 | 936 | |||
XM_017024195.1 | XP_016879684.1 | 4 | 936 | |||
XM_017024196.1 | XP_016879685.1 | 4 | 936 | |||
GenBank mRNA | AF044321.1 | - | 4 | 2411 |