Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257363.1 | NP_001244292.1 | 2 | 354 | ||
XM_005251708.3 | XP_005251765.1 | 2 | 346 | |||
XM_017014274.1 | XP_016869763.1 | 2 | 346 | |||
GenBank mRNA | AK313931.1 | - | 2 | 266 | ||
BI460020.1 | - | 2 | 330 |